  • Search Results
  • SERS-Active-Charged Microgels for Size- and Charge-Selective Molecular Analysis of Complex Biological Samples.
SERS-Active-Charged Microgels for Size- and Charge-Selective Molecular Analysis of Complex Biological Samples.
Small (Weinheim an der Bergstrasse, Germany) (2018-08-22)
Dong Jae Kim, Sung-Gyu Park, Dong-Ho Kim, Shin-Hyun Kim
ABSTRACT
Surface-enhanced Raman scattering (SERS) provides a dramatic increase of Raman intensity for molecules adsorbed on nanogap-rich metal nanostructures, serving as a promising tool for molecular analysis. However, surface contamination caused by protein adsorption and low surface concentration of small target molecules reduce the sensitivity, which severely restricts the use of SERS in many applications. Here, charged microgels containing agglomerates of gold nanoparticles (Au NPs) are designed using droplet-based microfluidics to provide a reliable SERS substrate with molecular selectivity and high sensitivity. The limiting mesh size of hydrogel enables the autonomous exclusion of large proteins and the charged matrix concentrates oppositely charged small molecules through electrostatic attraction. As nanogaps among Au NPs in the agglomerates enhance Raman intensity, Raman spectrum of the adsorbed molecules is selectively measured with high sensitivity in the absence of interruption from adhesive proteins. Therefore, the SERS-active-charged microgels can be used for direct analysis of pristine biological samples without the pretreatment steps of separation and concentration, which are commonly a prerequisite for Raman analysis. For the purpose of demonstration, a direct detection of fipronil sulfone with partial negative charges, a metabolite of toxic insecticide, dissolved in eggs using the positively charged microgels without any pretreatment of the samples, is shown.
Sigma-Aldrich
8-Hydroxypyrene-1,3,6-trisulfonic acid trisodium salt, ≥96%
Supelco
Phosmet, PESTANAL®, analytical standard
Sigma-Aldrich
Trimethoxy(octadecyl)silane, technical grade
Supelco
Fipronil sulfone, PESTANAL®, analytical standard