  • Search Results
  • Transcription Factors Drive Tet2-Mediated Enhancer Demethylation to Reprogram Cell Fate.
Transcription Factors Drive Tet2-Mediated Enhancer Demethylation to Reprogram Cell Fate.
Cell stem cell (2018-09-18)
Jose Luis Sardina, Samuel Collombet, Tian V Tian, Antonio Gómez, Bruno Di Stefano, Clara Berenguer, Justin Brumbaugh, Ralph Stadhouders, Carolina Segura-Morales, Marta Gut, Ivo G Gut, Simon Heath, Sergi Aranda, Luciano Di Croce, Konrad Hochedlinger, Denis Thieffry, Thomas Graf
ABSTRACT
Here, we report DNA methylation and hydroxymethylation dynamics at nucleotide resolution using C/EBPα-enhanced reprogramming of B cells into induced pluripotent cells (iPSCs). We observed successive waves of hydroxymethylation at enhancers, concomitant with a decrease in DNA methylation, suggesting active demethylation. Consistent with this finding, ablation of the DNA demethylase Tet2 almost completely abolishes reprogramming. C/EBPα, Klf4, and Tfcp2l1 each interact with Tet2 and recruit the enzyme to specific DNA sites. During reprogramming, some of these sites maintain high levels of 5hmC, and enhancers and promoters of key pluripotency factors become demethylated as early as 1 day after Yamanaka factor induction. Surprisingly, methylation changes precede chromatin opening in distinct chromatin regions, including Klf4 bound sites, revealing a pioneer factor activity associated with alternation in DNA methylation. Rapid changes in hydroxymethylation similar to those in B cells were also observed during compound-accelerated reprogramming of fibroblasts into iPSCs, highlighting the generality of our observations.
Sigma-Aldrich
DMOG, ≥98% (HPLC)
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Monoclonal Anti-β-Tubulin I antibody produced in mouse, clone SAP.4G5, ascites fluid
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)