US
EN
Applications
Products
Services
Support
Home
Search Results
Intensification of biopolymeric microparticles production by ultrasonic assisted atomization
Intensification of biopolymeric microparticles production by ultrasonic assisted atomization
Chemical Engineering and Processing (2009)
Barba AA, et al.