  • Search Results
  • Discovery of antivirulence agents against methicillin-resistant Staphylococcus aureus
Discovery of antivirulence agents against methicillin-resistant Staphylococcus aureus
Antimicrobial agents and chemotherapy (2013)
Khodaverdian V, et al.