  • Search Results
  • Autophagosomal Content Profiling Reveals an LC3C-Dependent Piecemeal Mitophagy Pathway.
Autophagosomal Content Profiling Reveals an LC3C-Dependent Piecemeal Mitophagy Pathway.
Molecular cell (2017-11-18)
François Le Guerroué, Franziska Eck, Jennifer Jung, Tatjana Starzetz, Michel Mittelbronn, Manuel Kaulich, Christian Behrends
ABSTRACT
Autophagy allows the degradation of cytosolic endogenous and exogenous material in the lysosome. Substrates are engulfed by double-membrane vesicles, coined autophagosomes, which subsequently fuse with lysosomes. Depending on the involvement of specific receptor proteins, autophagy occurs in a selective or nonselective manner. While this process is well understood at the level of bulky cargo such as mitochondria and bacteria, we know very little about individual proteins and protein complexes that are engulfed and degraded by autophagy. In contrast to the critical role of autophagy in balancing proteostasis, our current knowledge of the autophagic degradome is very limited. Here, we combined proximity labeling with quantitative proteomics to systematically map the protein inventory of autophagosomes. Using this strategy, we uncovered a basal, housekeeping mitophagy pathway that involves piecemeal degradation of mitochondrial proteins in a LC3C- and p62-dependent manner and contributes to mitochondrial homeostasis maintenance when cells rely on oxidative phosphorylation.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Millipore
ANTI-FLAG® M1 Agarose Affinity Gel
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.
Sigma-Aldrich
Anti-LC3C antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution