Molecular determinants for the tissue specificity of SERMs
Science (New York, N.Y.) (2002)
Shang, Y. and Brown, M.
Sigma-Aldrich
Anti-EF1α Antibody, clone CBP-KK1, clone CBP-KK1, Upstate®, from mouse
Pricing and availability is not currently available.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.