5-ht6 receptors.
Current Drug Targets. CNS and Neurological Disorders (2004)
Woolley, Marie L, et al.
Sigma-Aldrich
ChemiSCREEN Membrane Preparation Recombinant Human 5-HT6 Serotonin Receptor, Human 5-HT6 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Pricing and availability is not currently available.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.