  • Search Results
  • Anti-inflammatory and analgesic effects of a novel pyrazole derivative, FR140423.
Anti-inflammatory and analgesic effects of a novel pyrazole derivative, FR140423.
European journal of pharmacology (1999-02-13)
T Ochi, K Jobo-Magari, A Yonezawa, K Matsumori, T Fujii
ABSTRACT
The pharmacological profile of a novel and newly discovered non-steroidal anti-inflammatory and analgesic compound, 3-(difluoromethyl)-1-(4-methoxyphenyl)-5-[4-(methylsulfinyl)phenyl]pyraz ole (FR140423), was investigated. In recombinant human cyclooxygenase enzyme assays, the inhibition of prostaglandin E2 formation by FR140423 was 150 times more selective for cyclooxygenase-2 than cyclooxygenase-1. Oral administration of FR140423 dose dependently reduced carrageenin-induced paw edema and adjuvant arthritis. These effects were two- to three-fold more potent than those of indomethacin. Unlike indomethacin, FR140423 did not induce mucosal lesions in the stomach. FR140423 showed dose-dependent anti-hyperalgesic effects in the yeast-induced hyperalgesic model. This effect was five-fold more potent than that of indomethacin. Furthermore, FR140423 increased the pain threshold in non-inflamed paws and, unlike indomethacin, it produced an analgesic effect in the tail-flick test. These analgesic effects were blocked by the mu-opioid antagonist naloxone. These results suggest that FR140423, a selective cyclooxygenase-2 inhibitor, is a potent non-steroidal anti-inflammatory drug (NSAID) without gastrointestinal side effects and is a unique compound having morphine-like analgesic effects.
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.