  • Search Results
  • Structure of the elastase-cathepsin G inhibitor of the leech Hirudo medicinalis.
Structure of the elastase-cathepsin G inhibitor of the leech Hirudo medicinalis.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie (1980-12-01)
U Seemüller, M Eulitz, H Fritz, A Strobl
ABSTRACT
The leech Hirudo medicinalis contains three different groups of proteinase inhibitor proteins, the thrombin-specific hirudin, the bdellins directed against trypsin, plasmin and acrosin, and the eglins which were discovered only recently. We are interested in the eglins mainly for two reasons: (i) They form strong complexes with the granulocytic elastase and cathepsin G with Ki values close to 1 x 10(-10) mol/l. Due to this property they are potential candidates for the therapeutic treatment of various diseases. (ii) Although the eglins do not contain a disulfide bridge to stabilize the tertiary structure, they are highly resistant to denaturation by acidification and by heat as well as to proteolytic degradation.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.