  • Search Results
  • A large fragment approach to DNA synthesis: total synthesis of a gene for the protease inhibitor eglin c from the leech Hirudo medicinalis and its expression in E. coli.
A large fragment approach to DNA synthesis: total synthesis of a gene for the protease inhibitor eglin c from the leech Hirudo medicinalis and its expression in E. coli.
Nucleic acids research (1984-08-24)
H Rink, M Liersch, P Sieber, F Meyer
ABSTRACT
A DNA containing the coding sequence for the proteinase inhibitor protein, eglin c, from the leech Hirudo medicinalis has been obtained by enzymatic assembly of chemically synthesized DNA fragments. The synthetic gene consists of a 232 base-pair fragment containing initiation and termination codon signals with restriction enzyme recognition sites conveniently placed for cloning into a plasmid vector. Only six oligonucleotides from 34 to 61 bases in length, sharing pairwise stretches of complementary regions at their 3'-termini, were prepared by phosphotriester solid-phase synthesis. The oligomers were annealed pairwise and converted into double stranded DNA fragments by DNA polymerase I mediated repair synthesis. The fragments were assembled by ligation, and the synthetic gene was expressed in high yield in E. coli under the transcriptional control of the E. coli tryptophan promoter. The expression product was purified to homogeneity and was shown to have similar physicochemical and identical biological properties as the authentic protein isolated from the leech.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.