MilliporeSigma
Role for Med12 in regulation of Nanog and Nanog target genes.
The Journal of biological chemistry (2008-11-28)
Antonin V Tutter, Michael P Kowalski, Gretchen A Baltus, Vadim Iourgenko, Mark Labow, En Li, Shilpa Kadam
ABSTRACT
Nanog, Oct4, and Sox2 form the core of a transcription factor network that maintains embryonic stem cells in the pluripotent state in both humans and mice. These critical factors have been implicated as both positive and negative regulators of transcription, varying by promoter and differentiation state of the cell. The Mediator complex, a ubiquitous conserved complex of approximately 30 subunits, facilitates transcription by coordinating RNA polymerase II binding to target promoters via gene-specific activators and can be divided into several functional subcomplexes. Med12 is part of a subcomplex of four proteins associated with the core Mediator complex and has been found to function both in repressing and activating transcription when recruited to target promoters. We identified an interaction between Med12 and Nanog and present evidence of involvement of Med12 in regulation of Nanog function. Gene expression analysis of embryonic stem cells knocked down for Med12 showed a similarity to Nanog knockdown, with increased expression of Nanog-repressed targets and decreased expression of Nanog-activated targets. Using chromatin immunoprecipitation, we found that Med12 and Nanog co-occupied Nanog target promoters in embryonic stem cells and that Med12 dissociated from target promoters upon differentiation with kinetics similar to Nanog. Our results indicate that Nanog and Med12 function in concert to regulate Nanog target genes and identify a novel role for Med12 in embryonic stem cell regulation.
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Nanog Antibody, NT, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, serum, Upstate®