  • Search Results
  • Overproduction and secretion of a novel amino-terminal form of parathyroid hormone from a severe type of parathyroid hyperplasia in uremia.
Overproduction and secretion of a novel amino-terminal form of parathyroid hormone from a severe type of parathyroid hyperplasia in uremia.
Clinical journal of the American Society of Nephrology : CJASN (2007-08-21)
Toshio Arakawa, Pierre D'Amour, Louise Rousseau, Jean-Hugues Brossard, Makoto Sakai, Hiroomi Kasumoto, Naoya Igaki, Takeo Goto, Tom Cantor, Masafumi Fukagawa
ABSTRACT
Measurement of bioactive parathyroid hormone (PTH) is essential for optimal management of bone abnormalities in dialysis patients. This can be accomplished by PTH measurements using third-generation PTH assays, which detect more or less of the first six amino acids of the PTH structure. Such assays do not detect non-(1-84) PTH fragments, such as human PTH (7-84), which are recognized by the second-generation PTH assays that use a detection antibody that recognizes an epitope within the 13-34 region of the PTH structure. Therefore, third-generation PTH results are expected to be lower than those that are obtained with second-generation PTH assays. Rare exceptions to this rule have been reported for patients with severe primary hyperparathyroidism or parathyroid cancer. Sera and gland extracts were analyzed from a dialysis patient with high bone turnover disease and with surprising higher PTH levels by a third-generation assay than by a second-generation assay. This finding normalized after the surgical removal of an enlarged gland with a single nodule, an advanced type of nodular hyperplasia. HPLC fractionation of sera and gland extracts revealed the overproduction and secretion of a PTH molecule with an intact amino-terminus structure distinct from (1-84) PTH. This form of PTH was readily detectable by third-generation PTH assays but was poorly reactive in second-generation PTH assays. Therefore, parathyroid glands with advanced uremic nodular hyperplasia may overproduce and secrete a novel, biologically active form of PTH with an intact 1-6 region but a presumably modified 12-18 region required for the detection in second-generation PTH assays.
Sigma-Aldrich
Parathyroid Hormone Fragment 13-34 human, ≥95% (HPLC)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.