  • Search Results
  • Effects of intrauterine undernutrition on the expression of CYP3A23/3A1, PXR, CAR and HNF4alpha in neonate rats.
Effects of intrauterine undernutrition on the expression of CYP3A23/3A1, PXR, CAR and HNF4alpha in neonate rats.
Biopharmaceutics & drug disposition (2008-12-06)
Shaoqing Ni, Xiumin Wang, Jue Wang, Sijie Lu, Su Zeng, Zhengyan Zhao, Lushan Yu, Shuqing Chen
ABSTRACT
Cytochrome P-450 3A (CYP3A) together with its nuclear receptors plays a critical role in drug metabolism. The present study investigated the effects of undernutrition in utero on hepatic mRNA and protein expression of the enzyme CYP3A23/3A1 and nuclear receptors including pregnane X receptor (PXR; NR1I2), constitutive androstane receptor (CAR; NR1I3) and nuclear factor-4alpha (HNF4alpha; HNF4A) in neonatal rats. At gestational day 2, pregnant rats were randomly divided into two groups: nourished (fed ad libitum) and undernourished (50% of nourished group). The pups delivered by nourished rats were designated as the normal-birth-weight group (NBW, n=15) and those delivered by undernourished rats were designated as the low-birth-weight group (LBW, n=15). Hepatic mRNA expression was detected by quantitative real-time PCR and the corresponding protein expression was examined by immunohistochemistry (IHC). Compared with NBW pups, LBW pups tended to have lower mRNA expression levels of CYP3A23/3A1, PXR and CAR but higher levels of HNF4alpha. Only the CAR mRNA expression differences were significant (p<0.05). mRNA expression of CYP3A23/3A1 correlated with that of HNF4alpha in both the LBW(r=0.808, p=0.007) and NBW (r=0.452, p=0.012) groups. CYP3A23/3A1 and CAR protein expression differed between the two groups (CYP3A23/3A1, chi(2)=7.87, p=0.005; CAR, chi(2)=12.069, p=0.001). In conclusion, these findings suggest that undernutrition may influence the mRNA expression of CAR and protein expression of both CYP3A23/3A1 and CAR in neonatal rats. Since CYP3A23/3A1 and CAR are critically involved in drug metabolism, these results may have clinical implications for optimal medication in LBW children.
Sigma-Aldrich
Anti-Cytochrome P450 Enzyme CYP3A1 Antibody, serum, Chemicon®
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.