  • Search Results
  • Regulation of estrogen rapid signaling through arginine methylation by PRMT1.
Regulation of estrogen rapid signaling through arginine methylation by PRMT1.
Molecular cell (2008-07-29)
Muriel Le Romancer, Isabelle Treilleux, Nicolas Leconte, Yannis Robin-Lespinasse, Stéphanie Sentis, Katia Bouchekioua-Bouzaghou, Sophie Goddard, Stéphanie Gobert-Gosse, Laura Corbo
ABSTRACT
Evidence is emerging that estrogen receptor alpha (ERalpha) is central to the rapid transduction of estrogen signaling to the downstream kinase cascades; however, the mechanisms underlying this nongenomic function are not fully understood. Here we report a paradigm of ERalpha regulation through arginine methylation by PRMT1, which transiently methylates arginine 260 within the ERalpha DNA-binding domain. This methylation event is required for mediating the extranuclear function of the receptor by triggering its interaction with the p85 subunit of PI3K and Src. Furthermore, we find that the focal adhesion kinase (FAK), a Src substrate involved in the migration process, is also recruited in this complex. Our data indicate that the methylation of ERalpha is a physiological process occurring in the cytoplasm of normal and malignant epithelial breast cells and that ERalpha is hypermethylated in a subset of breast cancers.
Sigma-Aldrich
Anti-PRMT1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
4G10® Platinum, Anti-Phosphotyrosine Antibody (mouse monoclonal cocktail IgG2b), clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anti-FAK Antibody, Upstate®, from rabbit