  • Search Results
  • SPINKL, a Kazal-type serine protease inhibitor-like protein purified from mouse seminal vesicle fluid, is able to inhibit sperm capacitation.
SPINKL, a Kazal-type serine protease inhibitor-like protein purified from mouse seminal vesicle fluid, is able to inhibit sperm capacitation.
Reproduction (Cambridge, England) (2008-08-22)
Ming-Huei Lin, Robert Kuo-Kuang Lee, Yuh-Ming Hwu, Chung-Hao Lu, Shian-Ling Chu, Ying-Jie Chen, Wei-Chao Chang, Sheng-Hsiang Li
ABSTRACT
We report a secreted serine protease inhibitor Kazal-type-like (SPINKL) protein. The SPINKL protein was purified from mouse seminal vesicle secretions through a series of steps, including ion-exchange chromatography on a diethylaminoethyl-Sephacel column, gel filtration on a Sephadex G-75 column, and ion-exchange HPLC on a Q strong anion exchange column. Further analysis identified several SPINKL proteins with various N-linked carbohydrates. The SPINKL protein has six conserved cysteine residues that are nearly identical to those of members of the SPINK protein family. It was noted that the SPINKL protein showed no inhibitory activities against common serine proteases such as trypsin, chymotrypsin, subtilisin, or elastase. Spinkl mRNA and SPINKL proteins were found to be primarily expressed in seminal vesicles. Immunohistochemistry revealed that the SPINKL protein occurred in the luminal fluid and mucosal epithelium of the seminal vesicles and was regulated by testosterone. The SPINKL protein was able to bind onto sperm and enhance sperm motility. Also, it was able to suppress BSA-stimulated sperm capacitation and block sperm-oocyte interactions in vitro, suggesting that SPINKL may be a decapacitation factor.
Sigma-Aldrich
Diethylaminoethyl–Sephacel®, aqueous ethanol suspension, 40-160 μm (wet), exclusion limit ~1,000,000 Da
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.