  • Search Results
  • Glycothermal synthesis of assembled vanadium oxide nanostructures for gas sensing.
Glycothermal synthesis of assembled vanadium oxide nanostructures for gas sensing.
Journal of Nanoparticle Research (2012)
Fu H, et al.