  • Search Results
  • Structural Insights into the Process of GPCR-G Protein Complex Formation.
Structural Insights into the Process of GPCR-G Protein Complex Formation.
Cell (2019-05-14)
Xiangyu Liu, Xinyu Xu, Daniel Hilger, Philipp Aschauer, Johanna K S Tiemann, Yang Du, Hongtao Liu, Kunio Hirata, Xiaoou Sun, Ramon Guixà-González, Jesper M Mathiesen, Peter W Hildebrand, Brian K Kobilka
ABSTRACT
The crystal structure of the β2-adrenergic receptor (β2AR) bound to the G protein adenylyl cyclase stimulatory G protein (Gs) captured the complex in a nucleotide-free state (β2AR-Gsempty). Unfortunately, the β2AR-Gsempty complex does not provide a clear explanation for G protein coupling specificity. Evidence from several sources suggests the existence of a transient complex between the β2AR and GDP-bound Gs protein (β2AR-GsGDP) that may represent an intermediate on the way to the formation of β2AR-Gsempty and may contribute to coupling specificity. Here we present a structure of the β2AR in complex with the carboxyl terminal 14 amino acids from Gαs along with the structure of the GDP-bound Gs heterotrimer. These structures provide evidence for an alternate interaction between the β2AR and Gs that may represent an intermediate that contributes to Gs coupling specificity.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Millipore
FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Benzamidine hydrochloride hydrate, ≥99%
Millipore
ANTI-FLAG® M1 Agarose Affinity Gel
Sigma-Aldrich
Cholesteryl hemisuccinate
Sigma-Aldrich
1-Oleoyl-rac-glycerol, ≥99%
Sigma-Aldrich
2,2-Dimethyl-1,3-propanediol, 99%
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
Cholesteryl hemisuccinate tris salt, anionic detergent
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)