  • Search Results
  • The role of fluoroalcohols as counter anions for ion-pairing reversed-phase liquid chromatography/high-resolution electrospray ionization mass spectrometry analysis of oligonucleotides.
The role of fluoroalcohols as counter anions for ion-pairing reversed-phase liquid chromatography/high-resolution electrospray ionization mass spectrometry analysis of oligonucleotides.
Rapid communications in mass spectrometry : RCM (2019-01-23)
Rong Liu, Yanjiao Ruan, Zhongqiu Liu, Lingzhi Gong
ABSTRACT
Hexafluoroisopropanol (HFIP) has been widely used as a counter anion in the mobile phase for ion-pairing reversed-phase liquid chromatography/mass spectrometry (IP-RP-LC/MS) analysis of oligonucleotides. However, researchers are still searching for improvements to counter anions for LC/MS analysis of oligonucleotides. This study aimed to find alternatives to HFIP for analyzing oligonucleotides. The study was performed using an Agilent 1290 ultra-high-performance liquid chromatography (UHPLC) system coupled to an Agilent 6540 mass spectrometer by using an oligonucleotide BEH C18 column (100 × 2.1 mm, 1.7 μm). Buffer systems containing ion-pairing reagents (triethylamine, tripropylamine, hexylamine, dimethylbutylamine, diisopropylethylamine, N,N-dimethylcyclohexylamine, and octylamine) and fluoroalcohols (HFIP and hexafluoro-2-methyl-2-propanol (HFTP)) were compared chromatographically and mass spectrometrically. Results showed that HFTP has better desalting ability than HFIP, but both HFIP and HFTP have comparable effects on the separation of oligonucleotides sized from 10mer to 40mer for most of ion-pairing reagents, with the exception of triethylamine and N,N-dimethylcyclohexylamine, where HFIP performed better than HFTP. The choice of fluoroalcohols in IP-RP-LC/MS analysis of oligonucleotides depends on the type of ion-pairing reagents used in the mobile phase. As a guideline, we would recommend to use either HA-HFIP or HA-HFTP for small oligonucleotides, but TPA-HFTP for large oligonucleotides for IP-RP-LC/MS analysis of synthetic oligonucleotides.