  • Search Results
  • Efficient biosynthesis of (R)-3-amino-1-butanol by a novel (R)-selective transaminase from Actinobacteria sp.
Efficient biosynthesis of (R)-3-amino-1-butanol by a novel (R)-selective transaminase from Actinobacteria sp.
Journal of biotechnology (2019-03-12)
Xiao-Ling Tang, Nan-Nan Zhang, Guo-Yan Ye, Yu-Guo Zheng
ABSTRACT
(R)-3-amino-1-butanol is a key intermediate of Dolutegravir for the treatment of HIV/AIDS and its green and efficient biosynthesis has attracted a great deal of attention. Transaminases are currently used as promising biocatalyst for the synthesis of chiral amines. However, many transaminases have (S)-specificity and (R)-selective transaminases were less exploited and studied, making the production of (R)-amines remain challenging. In this study, a novel transaminase from Actinobacteria sp. (As-TA) was obtained and applied for the biosynthesis of (R)-3-amino-1-butanol by transferring the amino group from isopropylamine to 4-hydroxy-2-butanone. After optimization of the reaction condition and using a substrate fed-batch strategy, the conversion of 100, 200, 300, 400 and 500 mM 4-hydroxy-2-butanone reached 100%, 94.9%, 86.1%, 76.1% and 70.9%, respectively. (R)-3-amino-1-butanol with a maximum yield of 29.6 g/L and 99.9% e.e. value was obtained. This was the first time demonstrating the successful biosynthesis of (R)-3-amino-1-butanol with transaminase as biocatalyst and the obtained As-TA enriched the enzyme pool of transaminase with (R)-specificity.
Sigma-Aldrich
4-Hydroxy-2-butanone, 95%