  • Search Results
  • Influence of Triplet Diffusion on Lead Halide Perovskite-Sensitized Solid-State Upconversion.
Influence of Triplet Diffusion on Lead Halide Perovskite-Sensitized Solid-State Upconversion.
The journal of physical chemistry letters (2019-06-28)
Sarah Wieghold, Alexander S Bieber, Zachary A VanOrman, Lea Nienhaus
ABSTRACT
The emerging field of lead halide perovskite-sensitized triplet-triplet annihilation (TTA) in rubrene shows great promise in upconversion applications. By rapidly transferring single charge carriers instead of bound triplet states, perovskites enable a high triplet population in rubrene, yielding low Ith values. In this contribution, we investigate the role of the triplet population on the upconverted emission. Interestingly, two independent rates of TTA can be observed, as well as a sharp drop in the visible emission intensity over several seconds. This effect can be attributed to the triplet-density-based diffusion length: (i) at low triplet populations slow diffusion-mediated TTA yields singlets far from the interface and (ii) higher triplet populations lead to rapid TTA close to the perovskite/rubrene interface. Because of the proximity of the strongly absorbing perovskite, the singlet states created closer to the interface undergo stronger back-transfer to the perovskite film and therefore appear to exhibit a lower photoluminescence quantum yield.
Sigma-Aldrich
Rubrene, sublimed grade, 99.99% trace metals basis
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.