  • Search Results
  • Identifying cystogenic paracrine signaling molecules in cyst fluid of patients with polycystic kidney disease.
Identifying cystogenic paracrine signaling molecules in cyst fluid of patients with polycystic kidney disease.
American journal of physiology. Renal physiology (2018-11-08)
Annegien T Kenter, Sari E van Rossum-Fikkert, Mahdi Salih, Paul C M S Verhagen, Geert J L H van Leenders, Jeroen A A Demmers, Gert Jansen, Joost Gribnau, Robert Zietse, Ewout J Hoorn
ABSTRACT
In autosomal dominant polycystic kidney disease (ADPKD) paracrine signaling molecules in cyst fluid can induce proliferation and cystogenesis of neighboring renal epithelial cells. However, the identity of this cyst-inducing factor is still unknown. The aim of this study was to identify paracrine signaling proteins in cyst fluid using a 3D in vitro cystogenesis assay. We collected cyst fluid from 15 ADPKD patients who underwent kidney or liver resection (55 cysts from 13 nephrectomies, 5 cysts from 2 liver resections). For each sample, the ability to induce proliferation and cyst formation was tested using the cystogenesis assay (RPTEC/TERT1 cells in Matrigel with cyst fluid added for 14 days). Kidney cyst fluid induced proliferation and cyst growth of renal epithelial cells in a dose-dependent fashion. Liver cyst fluid also induced cystogenesis. Using size exclusion chromatography, 56 cyst fluid fractions were obtained of which only the fractions between 30 and 100 kDa showed cystogenic potential. Mass spectrometry analysis of samples that tested positive or negative in the assay identified 43 candidate cystogenic proteins. Gene ontology analysis showed an enrichment for proteins classified as enzymes, immunity proteins, receptors, and signaling proteins. A number of these proteins have previously been implicated in ADPKD, including secreted frizzled-related protein 4, S100A8, osteopontin, and cysteine rich with EGF-like domains 1. In conclusion, both kidney and liver cyst fluids contain paracrine signaling molecules that drive cyst formation. Using size exclusion chromatography and mass spectrometry, we procured a candidate list for future studies. Ultimately, cystogenic paracrine signaling molecules may be targeted to abrogate cystogenesis in ADPKD.
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine, ≥95% (HPLC), powder