  • Search Results
  • Basement membrane destruction by pancreatic stellate cells leads to local invasion in pancreatic ductal adenocarcinoma.
Basement membrane destruction by pancreatic stellate cells leads to local invasion in pancreatic ductal adenocarcinoma.
Cancer letters (2018-03-28)
Kazuhiro Koikawa, Kenoki Ohuchida, Yohei Ando, Shin Kibe, Hiromichi Nakayama, Shin Takesue, Sho Endo, Toshiya Abe, Takashi Okumura, Chika Iwamoto, Taiki Moriyama, Kohei Nakata, Yoshihiro Miyasaka, Takao Ohtsuka, Eishi Nagai, Kazuhiro Mizumoto, Makoto Hashizume, Masafumi Nakamura
ABSTRACT
Stroma invasion is an important step in pancreatic cancer progression. However, how pancreatic ductal adenocarcinoma (PDAC) with ductal structure invades the surrounding stroma has not been clear. Here, we elucidated the mechanism of stromal invasion of PDAC, using organoids. From resected PDAC specimens, we established human PDAC organoids, which developed ductal and basement membrane (BM) structures. When the organoids were co-cultured with pancreatic stellate cells (PSCs) in a collagen matrix, organoids lost their BM and ductal structures, and invaded collagen matrix more frequently than did mono-cultured organoids. Interestingly, direct contact by PSCs to PDAC organoids was observed before BM destruction. Matrix metalloproteinase (MMP) 2 or membrane type-1 MMP (MT1MMP) knockdown in PSCs significantly attenuated BM destruction by PSCs, and retained the ductal structures in organoids. Our results imply that direct contact by PSCs induces BM destruction and stromal invasion of PDAC via MMP2 which binds to MT1MMP on PSCs.
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)