  • Search Results
  • β-actin regulates a heterochromatin landscape essential for optimal induction of neuronal programs during direct reprograming.
β-actin regulates a heterochromatin landscape essential for optimal induction of neuronal programs during direct reprograming.
PLoS genetics (2018-12-18)
Xin Xie, Robertas Jankauskas, Aslam M A Mazari, Nizar Drou, Piergiorgio Percipalle
ABSTRACT
During neuronal development, β-actin serves an important role in growth cone mediated axon guidance. Consistent with this notion, in vivo ablation of the β-actin gene leads to abnormalities in the nervous system. However, whether β-actin is involved in the regulation of neuronal gene programs is not known. In this study, we directly reprogramed β-actin+/+ WT, β-actin+/- HET and β-actin-/- KO mouse embryonic fibroblast (MEFs) into chemically induced neurons (CiNeurons). Using RNA-seq analysis, we profiled the transcriptome changes among the CiNeurons. We discovered that induction of neuronal gene programs was impaired in KO CiNeurons in comparison to WT ones, whereas HET CiNeurons showed an intermediate levels of induction. ChIP-seq analysis of heterochromatin markers demonstrated that the impaired expression of neuronal gene programs correlated with the elevated H3K9 and H3K27 methylation levels at gene loci in β-actin deficient MEFs, which is linked to the loss of chromatin association of the BAF complex ATPase subunit Brg1. Together, our study shows that heterochromatin alteration in β-actin null MEFs impedes the induction of neuronal gene programs during direct reprograming. These findings are in line with the notion that H3K9Me3-based heterochromatin forms a major epigenetic barrier during cell fate change.
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
SAFC
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
SAFC
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and sodium bicarbonate, without L-glutamine and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
CHIR99021, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Fetal Bovine Serum, Research Grade, non-USA origin, sterile-filtered, suitable for cell culture