  • Search Results
  • Docosahexaenoic Acid Inhibits Proliferation of EoL-1 Leukemia Cells and Induces Cell Cycle Arrest and Cell Differentiation.
Docosahexaenoic Acid Inhibits Proliferation of EoL-1 Leukemia Cells and Induces Cell Cycle Arrest and Cell Differentiation.
Nutrients (2019-03-15)
Kalliopi Moustaka, Eirini Maleskou, Andromachi Lambrianidou, Stelios Papadopoulos, Marilena E Lekka, Theoni Trangas, Eirini Kitsiouli
ABSTRACT
Τhe effect of docosahexaenoic acid (DHA, an omega-3 polyunsaturated fatty acid) upon the proliferation of EoL-1 (Eosinophilic leukemia) cell line was assessed, while additional cellular events during the antiproliferative action were recorded. DHA inhibited EoL-1 cells growth dose-dependently by inducing growth arrest at G0/1 phase of the cell cycle. After DHA addition to the cells, the expression of MYC oncogene was decreased, PTAFR-mRNA overexpression was observed which was used as a marker of differentiation, and PLA2G4A-mRNA increase was recorded. The enzymatic activities of phospholipase A₂ (PLA₂), a group of hydrolytic enzymes, whose action precedes and leads to PAF biosynthesis through the remodeling pathway, as well as platelet activating factor acetylhydrolase (PAFAH) which hydrolyses and deactivates PAF, were also measured. DHA had an effect on the levels of both the intracellular and secreted activities of PLA₂ and PAFAH. The inflammatory cytokines IL-6 and TNF-α were also detected in high levels. In conclusion, DHA-induced EoL-1 cells differentiation was correlated with downregulation of MYC oncogene, overexpression of PTAFR and PLA2G4A-mRNAs, increase of the inflammatory cytokines production, and alteration of the enzymatic activities that regulate PAF levels. DHA is a natural substance and the understanding of its action on EoL-1 cells on molecular level could be useful in further investigation as a future therapeutic tool against F/P ⁺ hypereosinophilic syndrome.
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
cis-4,7,10,13,16,19-Docosahexaenoic acid, ≥98%
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)