  • Search Results
  • Macrophage stimulating protein preserves blood brain barrier integrity after intracerebral hemorrhage through recepteur d'origine nantais dependent GAB1/Src/β-catenin pathway activation in a mouse model.
Macrophage stimulating protein preserves blood brain barrier integrity after intracerebral hemorrhage through recepteur d'origine nantais dependent GAB1/Src/β-catenin pathway activation in a mouse model.
Journal of neurochemistry (2018-11-01)
Tai Lu, Zhong Wang, Sherchan Prativa, Yang Xu, Tian Wang, Yiting Zhang, Lingyan Yu, Ningbo Xu, Jiping Tang, Wanchun You, Gang Chen, John H Zhang
ABSTRACT
Blood brain barrier (BBB) disruption is an important contributor to brain edema and neurological deficits following intracerebral hemorrhage (ICH). Macrophage stimulating protein (MSP) is a hepatocyte growth factor-like protein that mediates its functions via activating receptor tyrosine kinase recepteur d'origine nantais (RON). Grb2-associated binder 1 (GAB1) is a docking protein that mediates downstream receptor signal transduction pathways. This study aimed to evaluate the role of MSP and RON activated signaling pathway in preserving BBB integrity after collagenase-induced ICH. ICH mice received recombinant human MSP (rhMSP) or rhMSP combined with siRNA knockdown of RON or GAB1. rhMSP was administered by intranasal route 1 h after ICH. Brain edema, neurobehavior, BBB tight junction protein expression, and BBB permeability were evaluated. The expression of endogenous MSP and p-RON was decreased after ICH. Exogenous rhMSP administration reduced brain edema, neurological deficits, BBB permeability, and increased the expression of tight junction proteins in ICH mice. rhMSP administration increased the expression of p-RON, p-GAB1, p-Src, nuclear β-catenin, and tight junction proteins after ICH. These effects were reversed with RON and GAB1 siRNA. We conclude that MSP activation of RON preserved BBB integrity via GAB-1/Src/β-catenin pathway, thereby reducing brain edema and neurological deficits after ICH in mice.
Sigma-Aldrich
Evans Blue, Dye content ≥75 %
Sigma-Aldrich
Phosphate buffered saline, BioPerformance Certified, pH 7.4
Sigma-Aldrich
Drabkin′s reagent, for the determination of hemoglobin
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Buprenorphine hydrochloride
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, ≥700 CDU/mg solid (CDU = collagen digestion units)