  • Search Results
  • Fluorescence lifetime-activated droplet sorting in microfluidic chip systems.
Fluorescence lifetime-activated droplet sorting in microfluidic chip systems.
Lab on a chip (2019-01-04)
Sadat Hasan, David Geissler, Konstantin Wink, Axel Hagen, Josef J Heiland, Detlev Belder
ABSTRACT
We present a highly efficient microfluidic fluorescence lifetime-activated droplet sorting (FLADS) approach as a novel technology for droplet manipulation in lab-on-a-chip devices. In a proof-of-concept study, we successfully applied the approach to sort droplets containing two different fluorescent compounds on the basis of their corresponding fluorescence lifetime. Towards this end, a technical set-up was developed enabling on-the-fly fluorescence lifetime determination of passing droplets. The herein developed LabVIEW program enabled fast triggering of a downstream dielectrophoretic force sorting functionality depending on average fluorescence lifetimes of individual droplets. The approach worked reliably at individual substrate concentrations from 1 nM to 1 mM. This not only allowed reliable sorting of droplets containing species with different fluorescence lifetimes but also enabled differentiation of mixtures in individual droplets.