  • Search Results
  • Contribution of p38 MAPK Pathway to Norcantharidin-Induced Programmed Cell Death in Human Oral Squamous Cell Carcinoma.
Contribution of p38 MAPK Pathway to Norcantharidin-Induced Programmed Cell Death in Human Oral Squamous Cell Carcinoma.
International journal of molecular sciences (2019-07-19)
Chi-Hyun Ahn, Kyoung-Ok Hong, Bohwan Jin, WonWoo Lee, Yun Chan Jung, Hakmo Lee, Ji-Ae Shin, Sung-Dae Cho, Seong Doo Hong
ABSTRACT
Norcantharidin (NCTD), a demethylated analog of cantharidin isolated from blister beetles, has been used as a promising anticancer agent; however, the underlying function of NCTD against human oral squamous cell carcinoma (OSCC) has not been fully understood. Here, this study was aimed to investigate the apoptotic effect and molecular targets of NCTD in human OSCC in vitro and in vivo. The anticancer effects of NCTD and its related molecular mechanisms were evaluated by trypan blue exclusion assay, live/dead assay, western blotting, 4-6-Diamidino-2-Phenylindole (DAPI) staining, flow cytometric analysis, Terminal Deoxynucleotidyl Transferase dUTP Nick end Labeling (TUNEL) assay, and immunohistochemistry. NCTD significantly inhibited cell growth and increased the number of dead cells in HSC-3 and HN22 cell lines. It induced the following apoptotic phenomena: (1) the cleavages of poly (ADP-ribose) polymerase and casepase-3; (2) increase in apoptotic morphological changes (nuclear condensation and fragmentation); (3) increase in annexin V-positive cells or sub-G1 population of cells. NCTD significantly activated the p38 mitogen-activated protein kinase (MAPK) pathway but inactivated the signal transducer and activator of transcription (STAT)3 pathway. A p38 MAPK inhibitor (SB203580) partially attenuated NCTD-induced programmed cell death (apoptosis) in both cell lines, whereas ectopic overexpression of STAT3 did not affect it. NCTD strongly suppressed tumor growth in the tumor xenograft bearing HSC-3 cells, and the number of TUNEL-positive cells increased in NCTD-treated tumor tissues. In addition, NCTD did not cause any histopathological changes in the liver nor the kidney. NCTD induced programmed cell death via the activation of p38 MAPK in OSCC. Therefore, these results suggest that NCTD could be a potential anticancer drug candidate for the treatment of OSCC.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)