  • Search Results
  • Human Fetal TNF-α-Cytokine-Producing CD4+ Effector Memory T Cells Promote Intestinal Development and Mediate Inflammation Early in Life.
Human Fetal TNF-α-Cytokine-Producing CD4+ Effector Memory T Cells Promote Intestinal Development and Mediate Inflammation Early in Life.
Immunity (2019-02-17)
Renée R C E Schreurs, Martin E Baumdick, Adrian F Sagebiel, Max Kaufmann, Michal Mokry, Paul L Klarenbeek, Nicola Schaltenberg, Fenja L Steinert, Jorik M van Rijn, Agata Drewniak, Sarah-May M L The, Roel Bakx, Joep P M Derikx, Niek de Vries, Willemijn E Corpeleijn, Steven T Pals, Nicola Gagliani, Manuel A Friese, Sabine Middendorp, Edward E S Nieuwenhuis, Konrad Reinshagen, Teunis B H Geijtenbeek, Johannes B van Goudoever, Madeleine J Bunders
ABSTRACT
Although the fetal immune system is considered tolerogenic, preterm infants can suffer from severe intestinal inflammation, including necrotizing enterocolitis (NEC). Here, we demonstrate that human fetal intestines predominantly contain tumor necrosis factor-α (TNF-α)+CD4+CD69+ T effector memory (Tem) cells. Single-cell RNA sequencing of fetal intestinal CD4+ T cells showed a T helper 1 phenotype and expression of genes mediating epithelial growth and cell cycling. Organoid co-cultures revealed a dose-dependent, TNF-α-mediated effect of fetal intestinal CD4+ T cells on intestinal stem cell (ISC) development, in which low T cell numbers supported epithelial development, whereas high numbers abrogated ISC proliferation. CD4+ Tem cell frequencies were higher in inflamed intestines from preterm infants with NEC than in healthy infant intestines and showed enhanced TNF signaling. These findings reveal a distinct population of TNF-α-producing CD4+ T cells that promote mucosal development in fetal intestines but can also mediate inflammation upon preterm birth.
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Calcium chloride, anhydrous, granular, ≤7.0 mm, ≥93.0%
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Ionomycin calcium salt from Streptomyces conglobatus, powder, ≥98% (HPLC)
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Percoll®, Cytiva 17-0891-01, pack of 1 L
Sigma-Aldrich
SB 202190, ≥98% (HPLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
[Leu15]-Gastrin I human, ≥95% (HPLC)
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0%