  • Search Results
  • Trodusquemine enhances Aβ42 aggregation but suppresses its toxicity by displacing oligomers from cell membranes.
Trodusquemine enhances Aβ42 aggregation but suppresses its toxicity by displacing oligomers from cell membranes.
Nature communications (2019-01-16)
Ryan Limbocker, Sean Chia, Francesco S Ruggeri, Michele Perni, Roberta Cascella, Gabriella T Heller, Georg Meisl, Benedetta Mannini, Johnny Habchi, Thomas C T Michaels, Pavan K Challa, Minkoo Ahn, Samuel T Casford, Nilumi Fernando, Catherine K Xu, Nina D Kloss, Samuel I A Cohen, Janet R Kumita, Cristina Cecchi, Michael Zasloff, Sara Linse, Tuomas P J Knowles, Fabrizio Chiti, Michele Vendruscolo, Christopher M Dobson
ABSTRACT
Transient oligomeric species formed during the aggregation process of the 42-residue form of the amyloid-β peptide (Aβ42) are key pathogenic agents in Alzheimer's disease (AD). To investigate the relationship between Aβ42 aggregation and its cytotoxicity and the influence of a potential drug on both phenomena, we have studied the effects of trodusquemine. This aminosterol enhances the rate of aggregation by promoting monomer-dependent secondary nucleation, but significantly reduces the toxicity of the resulting oligomers to neuroblastoma cells by inhibiting their binding to the cellular membranes. When administered to a C. elegans model of AD, we again observe an increase in aggregate formation alongside the suppression of Aβ42-induced toxicity. In addition to oligomer displacement, the reduced toxicity could also point towards an increased rate of conversion of oligomers to less toxic fibrils. The ability of a small molecule to reduce the toxicity of oligomeric species represents a potential therapeutic strategy against AD.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-Amyloid β Antibody, clone W0-2, clone WO2, from mouse
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.