MilliporeSigma
  • Fibroblasts Mobilize Tumor Cell Glycogen to Promote Proliferation and Metastasis.
Fibroblasts Mobilize Tumor Cell Glycogen to Promote Proliferation and Metastasis.
Cell metabolism (2018-09-04)
Marion Curtis, Hilary A Kenny, Bradley Ashcroft, Abir Mukherjee, Alyssa Johnson, Yilin Zhang, Ynes Helou, Raquel Batlle, Xiaojing Liu, Nuria Gutierrez, Xia Gao, S Diane Yamada, Ricardo Lastra, Anthony Montag, Nagib Ahsan, Jason W Locasale, Arthur R Salomon, Angel R Nebreda, Ernst Lengyel
ABSTRACT
Successful metastasis requires the co-evolution of stromal and cancer cells. We used stable isotope labeling of amino acids in cell culture coupled with quantitative, label-free phosphoproteomics to study the bidirectional signaling in ovarian cancer cells and human-derived, cancer-associated fibroblasts (CAFs) after co-culture. In cancer cells, the interaction with CAFs supported glycogenolysis under normoxic conditions and induced phosphorylation and activation of phosphoglucomutase 1, an enzyme involved in glycogen metabolism. Glycogen was funneled into glycolysis, leading to increased proliferation, invasion, and metastasis of cancer cells co-cultured with human CAFs. Glycogen mobilization in cancer cells was dependent on p38α MAPK activation in CAFs. In vivo, deletion of p38α in CAFs and glycogen phosphorylase inhibition in cancer cells reduced metastasis, suggesting that glycogen is an energy source used by cancer cells to facilitate metastatic tumor growth.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
NSC23766 trihydrochloride, ≥97% (HPLC)
Pricing and availability is not currently available.