  • Search Results
  • Mechanisms of PKC-Mediated Enhancement of HIF-1α Activity and its Inhibition by Vitamin K2 in Hepatocellular Carcinoma Cells.
Mechanisms of PKC-Mediated Enhancement of HIF-1α Activity and its Inhibition by Vitamin K2 in Hepatocellular Carcinoma Cells.
International journal of molecular sciences (2019-03-01)
Jinghe Xia, Iwata Ozaki, Sachiko Matsuhashi, Takuya Kuwashiro, Hirokazu Takahashi, Keizo Anzai, Toshihiko Mizuta
ABSTRACT
Hypoxia-inducible factor 1 (HIF-1) plays important roles in cancer cell biology. HIF-1α is reportedly activated by several factors, including protein kinase C (PKC), in addition to hypoxia. We investigated the role of PKC isoforms and the effects of vitamin K2 (VK2) in the activation process of HIF-1α. Human hepatocellular carcinoma (HCC)-derived Huh7 cells were cultured under normoxic and hypoxic (1% O₂) conditions with or without the PKC stimulator TPA. The expression, transcriptional activity and nuclear translocation of HIF-1α were examined under treatment with PKC inhibitors, siRNAs against each PKC isoform and VK2. Hypoxia increased the expression and activity of HIF-1α. TPA increased the HIF-1α activity several times under both normoxic and hypoxic conditions. PKC-δ siRNA-mediated knockdown, PKC-δ inhibitor (rottlerin) and pan-PKC inhibitor (Ro-31-8425) suppressed the expression and transcriptional activity of HIF-1α. VK2 significantly inhibited the TPA-induced HIF-1α transcriptional activity and suppressed the expression and nuclear translocation of HIF-1α induced by TPA without altering the HIF-1α mRNA levels. These data indicate that PKC-δ enhances the HIF-1α transcriptional activity by increasing the nuclear translocation, and that VK2 might suppress the HIF-1α activation through the inhibition of PKC in HCC cells.
Sigma-Aldrich
Tris(2-pyridylmethyl)amine, 98%