  • Search Results
  • Molecular-Level Profiling of Human Serum Transferrin Protein through Assessment of Nanopore-Based Electrical and Chemical Responsiveness.
Molecular-Level Profiling of Human Serum Transferrin Protein through Assessment of Nanopore-Based Electrical and Chemical Responsiveness.
ACS nano (2019-03-08)
Jugal Saharia, Y M Nuwan D Y Bandara, Gaurav Goyal, Jung Soo Lee, Buddini Iroshika Karawdeniya, Min Jun Kim
ABSTRACT
In this study, we investigated the voltage and pH responsiveness of human serum transferrin (hSTf) protein using silicon nitride (Si xN y) nanopores. The Fe(III)-rich holo form of hSTf was dominant when pH > pI, while the Fe(III)-free apo form was dominant when pH < pI. The translocations of hSTf were purely in an electrophoretic sense, thus depended on its pI and the solution pH. With increasing voltage, voltage driven protein unfolding became prominent which was indicated by the trends associated with change in conductance, due to hSTf translocation, and in the excluded electrolyte volume. Additionally, analysis of the translocation events of the pure apo form of hSTf showed a clear difference in the event population compared to that of the holo form. The results obtained demonstrate the successful application of nanopore devices to distinguish between the holo and apo forms of hSTf in a mixture and to analyze its folding and unfolding phenomenon over a range of pH and applied voltages.
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
holo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
apo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
HEPES potassium salt, ≥99.5% (titration)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.