  • Search Results
  • Derivation of iPSC lines from two patients with familial Alzheimer's disease from India.
Derivation of iPSC lines from two patients with familial Alzheimer's disease from India.
Stem cell research (2019-01-04)
Ashaq H Najar, K M Sneha, Aparna Ashok, Swathy Babu, Anand G Subramaniam, Ramkrishnan Kannan, Biju Viswanath, Meera Purushottam, Mathew Varghese, Suhel Parvez, Mitradas M Panicker, Odity Mukherjee, Sanjeev Jain
ABSTRACT
The current prevalence of diagnosable dementia in India is 1% of people over 60 years (~3.7 million people), but is estimated to increase significantly, as ~15% world's aged population (>65 years) would be resident here by 2020 (Shah et al., 2016). While several mutations that pose a familial risk have been identified, the ethnic background may influence disease susceptibility, clinical presentation and treatment response. In this study, we report a detailed characterization of two representative HiPSC lines from a well-characterized dementia cohort from India. Availability of these lines, and associated molecular and clinical information, would be useful in the detailed exploration of the genomic contribution(s) to AD.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate, ≥95%
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)