  • Search Results
  • Spatial positioning of EB family proteins at microtubule tips involves distinct nucleotide-dependent binding properties.
Spatial positioning of EB family proteins at microtubule tips involves distinct nucleotide-dependent binding properties.
Journal of cell science (2018-09-29)
Daniel Roth, Benjamin P Fitton, Nikola P Chmel, Natalia Wasiluk, Anne Straube
ABSTRACT
EB proteins track the ends of growing microtubules and regulate microtubule dynamics both directly and by acting as the hub of the tip-tracking network. Mammalian cells express cell type-specific combinations of three EB proteins with different cellular roles. Here, we reconstitute EB1, EB2 and EB3 tip tracking in vitro We find that all three EBs show rapid exchange at the microtubule tip and that their signal correlates to the microtubule assembly rate. However, the three signals differ in their maxima and position from the microtubule tip. Using microtubules built with nucleotide analogues and site-directed mutagenesis, we show that EB2 prefers binding to microtubule lattices containing a 1:1 mixture of different nucleotides and its distinct binding specificity is conferred by amino acid substitutions at the right-hand-side interface of the EB microtubule-binding domain with tubulin. Our data are consistent with the model that all three EB paralogues sense the nucleotide state of both β-tubulins flanking their binding site. Their different profile of preferred binding sites contributes to occupying spatially distinct domains at the temporally evolving microtubule tip structure.
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered