G3BP1 promotes DNA binding and activation of cGAS.
Nature immunology (2018-12-05)
Zhao-Shan Liu, Hong Cai, Wen Xue, Miao Wang, Tian Xia, Wan-Jin Li, Jia-Qing Xing, Ming Zhao, Yi-Jiao Huang, Shuai Chen, Sheng-Ming Wu, Xinzheng Wang, Xin Liu, Xue Pang, Zi-Yu Zhang, Tingting Li, Jiang Dai, Fangting Dong, Qing Xia, Ai-Ling Li, Tao Zhou, Zheng-Gang Liu, Xue-Min Zhang, Tao Li
ABSTRACT
Cyclic GMP-AMP synthase (cGAS) is a key sensor responsible for cytosolic DNA detection. Here we report that GTPase-activating protein SH3 domain-binding protein 1 (G3BP1) is critical for DNA sensing and efficient activation of cGAS. G3BP1 enhanced DNA binding of cGAS by promoting the formation of large cGAS complexes. G3BP1 deficiency led to inefficient DNA binding by cGAS and inhibited cGAS-dependent interferon (IFN) production. The G3BP1 inhibitor epigallocatechin gallate (EGCG) disrupted existing G3BP1-cGAS complexes and inhibited DNA-triggered cGAS activation, thereby blocking DNA-induced IFN production both in vivo and in vitro. EGCG administration blunted self DNA-induced autoinflammatory responses in an Aicardi-Goutières syndrome (AGS) mouse model and reduced IFN-stimulated gene expression in cells from a patient with AGS. Thus, our study reveals that G3BP1 physically interacts with and primes cGAS for efficient activation. Furthermore, EGCG-mediated inhibition of G3BP1 provides a potential treatment for cGAS-related autoimmune diseases.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Sodium (meta)arsenite, ≥90%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
TRI Reagent®, for DNA, RNA and protein isolation
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Anti-MB21D1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-MB21D1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL3605, purified immunoglobulin, buffered aqueous glycerol solution