  • Search Results
  • Untangling hidden nutrient dynamics: rapid ammonium cycling and single-cell ammonium assimilation in marine plankton communities.
Untangling hidden nutrient dynamics: rapid ammonium cycling and single-cell ammonium assimilation in marine plankton communities.
The ISME journal (2019-03-27)
Isabell Klawonn, Stefano Bonaglia, Martin J Whitehouse, Sten Littmann, Daniela Tienken, Marcel M M Kuypers, Volker Brüchert, Helle Ploug
ABSTRACT
Ammonium is a central nutrient in aquatic systems. Yet, cell-specific ammonium assimilation among diverse functional plankton is poorly documented in field communities. Combining stable-isotope incubations (15N-ammonium, 15N2 and 13C-bicarbonate) with secondary-ion mass spectrometry, we quantified bulk ammonium dynamics, N2-fixation and carbon (C) fixation, as well as single-cell ammonium assimilation and C-fixation within plankton communities in nitrogen (N)-depleted surface waters during summer in the Baltic Sea. Ammonium production resulted from regenerated (≥91%) and new production (N2-fixation, ≤9%), supporting primary production by 78-97 and 2-16%, respectively. Ammonium was produced and consumed at balanced rates, and rapidly recycled within 1 h, as shown previously, facilitating an efficient ammonium transfer within plankton communities. N2-fixing cyanobacteria poorly assimilated ammonium, whereas heterotrophic bacteria and picocyanobacteria accounted for its highest consumption (~20 and ~20-40%, respectively). Surprisingly, ammonium assimilation and C-fixation were similarly fast for picocyanobacteria (non-N2-fixing Synechococcus) and large diatoms (Chaetoceros). Yet, the population biomass was high for Synechococcus but low for Chaetoceros. Hence, autotrophic picocyanobacteria and heterotrophic bacteria, with their high single-cell assimilation rates and dominating population biomass, competed for the same nutrient source and drove rapid ammonium dynamics in N-depleted marine waters.
Sigma-Aldrich
Ammonium-15N chloride, ≥98 atom % 15N, ≥99% (CP)
Sigma-Aldrich
Sodium bicarbonate-13C, 98 atom % 13C, 99% (CP)
Sigma-Aldrich
Nitrogen-15N2, 98 atom % 15N
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.