  • Search Results
  • Selenium Drives a Transcriptional Adaptive Program to Block Ferroptosis and Treat Stroke.
Selenium Drives a Transcriptional Adaptive Program to Block Ferroptosis and Treat Stroke.
Cell (2019-05-06)
Ishraq Alim, Joseph T Caulfield, Yingxin Chen, Vivek Swarup, Daniel H Geschwind, Elena Ivanova, Javier Seravalli, Youxi Ai, Lauren H Sansing, Emma J Ste Marie, Robert J Hondal, Sushmita Mukherjee, John W Cave, Botir T Sagdullaev, Saravanan S Karuppagounder, Rajiv R Ratan
ABSTRACT
Ferroptosis, a non-apoptotic form of programmed cell death, is triggered by oxidative stress in cancer, heat stress in plants, and hemorrhagic stroke. A homeostatic transcriptional response to ferroptotic stimuli is unknown. We show that neurons respond to ferroptotic stimuli by induction of selenoproteins, including antioxidant glutathione peroxidase 4 (GPX4). Pharmacological selenium (Se) augments GPX4 and other genes in this transcriptional program, the selenome, via coordinated activation of the transcription factors TFAP2c and Sp1 to protect neurons. Remarkably, a single dose of Se delivered into the brain drives antioxidant GPX4 expression, protects neurons, and improves behavior in a hemorrhagic stroke model. Altogether, we show that pharmacological Se supplementation effectively inhibits GPX4-dependent ferroptotic death as well as cell death induced by excitotoxicity or ER stress, which are GPX4 independent. Systemic administration of a brain-penetrant selenopeptide activates homeostatic transcription to inhibit cell death and improves function when delivered after hemorrhagic or ischemic stroke.
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Trimethoprim, ≥98% (HPLC)
Sigma-Aldrich
Erastin, ≥98% (HPLC)
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
(+)-MK-801 hydrogen maleate, powder
Sigma-Aldrich
LB Broth (Lennox), EZMix powder microbial growth medium
Sigma-Aldrich
2-Chlorotrityl chloride, ≥97.0% (AT)
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxid hexafluorophosphate, ChemDose tablets, Loading: 0.02mmol per tablet
Sigma-Aldrich
L-Homocysteic acid
Sigma-Aldrich
EZ-Magna ChIP® A - Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A beads. Control primers included.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)