  • Search Results
  • miR-552-5p facilitates osteosarcoma cell proliferation and metastasis by targeting WIF1.
miR-552-5p facilitates osteosarcoma cell proliferation and metastasis by targeting WIF1.
Experimental and therapeutic medicine (2019-04-17)
Wei Cai, Yong Xu, Jian Yin, Wenshan Zuo, Zhen Su
ABSTRACT
miR-552 promotes tumor growth and metastasis in colorectal cancer. However, the function of miR-552 in osteosarcoma remains unclear. The current study investigated the role and mechanism of miR-552-5p in the proliferation, migration and invasion of osteosarcoma cells. miR-552-5p was significantly upregulated in osteosarcoma tissues and cell lines compared with adjacent normal tissues and normal osteoblast cells. Knockdown of miR-552-5p significantly reduced the proliferation of MG63 and U2OS cells, and inhibited cell migration and invasion. Wnt inhibitory factor 1 (WIF1) was the direct target gene of miR-552-5p in osteosarcoma cells. Overexpression of miR-552-5p markedly decreased the expression of WIF1 in MG63 and U2OS cells. A negative association was identified between the expression levels of miR-552-5p and WIF1 in osteosarcoma tissues. Furthermore, the expression of WIF1 was downregulated in osteosarcoma tissues and cell lines. Finally, knockdown of WIF1 in MG63 and U2OS cells treated with miR-552-5p inhibitors rescued their ability to proliferate, migrate and invade. Overall, the results indicated that miR-552-5p promoted osteosarcoma development and progression by inhibiting WIF1. Therefore, miR-552-5p may serve as a new therapeutic target for treatment of patients with osteosarcoma.
Sigma-Aldrich
RNase A solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.