  • Search Results
  • Impact of repeated lipopolysaccharide administration on ovarian signaling during the follicular phase of the estrous cycle in post-pubertal pigs.
Impact of repeated lipopolysaccharide administration on ovarian signaling during the follicular phase of the estrous cycle in post-pubertal pigs.
Journal of animal science (2018-07-10)
Katie L Bidne, Sara S Kvidera, Jason W Ross, Lance H Baumgard, Aileen F Keating
ABSTRACT
Increased circulating lipopolysaccharide (LPS) results from heat stress (HS) and bacterial infection, both of which are associated with reduced female fertility. Specific effects of low-level, repeated LPS exposure on the ovary are unclear, as many studies utilize a bolus model and/or high dosage paradigm. To better understand the effects of chronic LPS exposure on ovarian signaling and function, post-pubertal gilts (n = 20) were orally administered altrenogest for 14 d to synchronize the beginning of the follicular phase of the ovarian cycle. For 5 d after synchronization, gilts (163 ± 3 kg) received IV administration of LPS (0.1 µg/kg BW, n = 10) or saline (CT, n = 10) 4× daily. Blood samples were obtained on days 1, 3, and 5 of LPS treatment. Follicular fluid was aspirated from dominant follicles on day 5, and whole ovarian homogenate was used for transcript and protein abundance analysis via quantitative real-time PCR and western blotting, respectively. There were no treatment differences detected in rectal temperature on any day (P ≥ 0.5). Administering LPS increased plasma insulin (P < 0.01), LPS-binding protein (LBP; P < 0.01), and glucose (P = 0.08) on day 1, but no treatment differences thereafter were observed (P = 0.66). There were no treatment differences in follicular fluid concentration of LBP or 17β-estradiol (P = 0.42). Gilts treated with LPS had increased abundance of ovarian TLR4 protein (P = 0.01), but protein kinase B (AKT) and phosphorylated AKT (pAKT) were unchanged and no effect of LPS on components of the phosphatidylinositol 3 kinase (PI3K) pathway were observed. There was no impact of LPS on ovarian abundance of STAR or CYP19A1, nor ESR1, LDLR, CYP19A1, CYP17A1, or 3BHSD. In conclusion, repeated, low-level LPS administration alters inflammatory but not steroidogenic or PI3K signaling in follicular phase gilt ovaries.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ponceau S, BioReagent, suitable for electrophoresis
Sigma-Aldrich
HEPES solution, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.