  • Search Results
  • A method using angiotensin converting enzyme immobilized on magnetic beads for inhibitor screening.
A method using angiotensin converting enzyme immobilized on magnetic beads for inhibitor screening.
Journal of pharmaceutical and biomedical analysis (2018-11-06)
Weiwei Tang, Bingjie Jia, Jie Zhou, Jing Liu, Jiancheng Wang, Dingyuan Ma, Ping Li, Jun Chen
ABSTRACT
Angiotensin converting enzyme (ACE), fusing with FLAG tag, was overexpressed in human embryonic kidney 293T cells. This recombinant FLAG-tagged ACE was immobilized on anti-FLAG antibody coated magnetic beads by affinity method in crude cell lysate for the first time. The enzyme-immobilized magnetic beads (ACE-MB), without further cleavage procedure, were used directly to establish a cost-effective and reliable method for screening ACE inhibitors by coupling with fluorescence detection. The enzymatic activity of the ACE-MB was validated based on its Michaelian kinetic behavior towards hippuryl-histidyl-leucine by UHPLC-MS/MS method firstly. Then, several conditions were optimized including amount of magnetic beads, incubation temperature and time in the procedure of ACE immobilization and amount of ACE-MB in the microplate operation. Moreover, this screening assay was validated with Z' factors between 0.71 and 0.81 using four known ACE inhibitors (captopril, lisinopril, fosinopril and fosinoprilat). The developed method was applied for the screening of ACE inhibitors from a small compound library of 45 natural products. As a result, epiberberine and fangchinoline with certain ACE inhibitory activities were screened out in the assay and validated. The results demonstrate the usefulness of this screening method using ACE immobilized on magnetic beads and the advantage of great efficiency with respect to both time and reagents for screening ACE inhibitors.
Sigma-Aldrich
Hippuric acid, 98%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.