  • Search Results
  • Functional importance of the oligomer formation of the cyanobacterial H+ pump Gloeobacter rhodopsin.
Functional importance of the oligomer formation of the cyanobacterial H+ pump Gloeobacter rhodopsin.
Scientific reports (2019-07-26)
Azusa Iizuka, Kousuke Kajimoto, Tomotsumi Fujisawa, Takashi Tsukamoto, Tomoyasu Aizawa, Naoki Kamo, Kwang-Hwan Jung, Masashi Unno, Makoto Demura, Takashi Kikukawa
ABSTRACT
Many microbial rhodopsins self-oligomerize, but the functional consequences of oligomerization have not been well clarified. We examined the effects of oligomerization of a H+ pump, Gloeobacter rhodopsin (GR), by using nanodisc containing trimeric and monomeric GR. The monomerization did not appear to affect the unphotolyzed GR. However, we found a significant impact on the photoreaction: The monomeric GR showed faint M intermediate formation and negligible H+ transfer reactions. These changes reflected the elevated pKa of the Asp121 residue, whose deprotonation is a prerequisite for the functional photoreaction. Here, we focused on His87, which is a neighboring residue of Asp121 and conserved among eubacterial H+ pumps but replaced by Met in an archaeal H+ pump. We found that the H87M mutation removes the "monomerization effects": Even in the monomeric state, H87M contained the deprotonated Asp121 and showed both M formation and distinct H+ transfer reactions. Thus, for wild-type GR, monomerization probably strengthens the Asp121-His87 interaction and thereby elevates the pKa of Asp121 residue. This strong interaction might occur due to the loosened protein structure and/or the disruption of the interprotomer interaction of His87. Thus, the trimeric assembly of GR enables light-induced H+ transfer reactions through adjusting the positions of key residues.