  • Search Results
  • Impact of post-procedural minimal stent area on 2-year clinical outcomes in the SYNTAX II trial.
Impact of post-procedural minimal stent area on 2-year clinical outcomes in the SYNTAX II trial.
Catheterization and cardiovascular interventions : official journal of the Society for Cardiac Angiography & Interventions (2019-02-01)
Yuki Katagiri, Giovanni Luigi De Maria, Norihiro Kogame, Ply Chichareon, Kuniaki Takahashi, Chun Chin Chang, Rodrigo Modolo, Simon Walsh, Manel Sabate, Justin Davies, Maciej Lesiak, Raul Moreno, Ignacio Cruz-Gonzalez, Nick E J West, Jan J Piek, Joanna J Wykrzykowska, Vasim Farooq, Javier Escaned, Adrian P Banning, Yoshinobu Onuma, Patrick W Serruys
ABSTRACT
To investigate the impact of minimal stent area (MSA) evaluated by post-procedural intravascular ultrasound (IVUS) on clinical outcomes after contemporary PCI in patients with three-vessel disease (TVD). The impact of post-procedural MSA on clinical outcomes has not yet been extensively studied in patients with TVD. The SYNTAX II study is a multicenter, all-comers, open-label, single arm study that investigated the impact of a state-of-the-art PCI strategy on clinical outcomes in patients with TVD (454 patients with 1,559 lesions). The relationships between post-procedural MSA and lesion-level outcomes at 2 years were investigated. Clinical events adjudicated per patient by clinical event committee were assessed per lesion. Lesion-oriented composite endpoint (LOCE) was defined as the composite of cardiac death, target-vessel myocardial infarction, and ischemia-driven target lesion revascularization. Eight hundred and nineteen lesions with post-procedural MSA available in 367 patients were included in the analysis. The post-procedural MSA per lesion was divided into terciles (smallest tercile: ≤5.0 mm2 , intermediate tercile: 5.0-6.7 mm2 , and largest tercile: >6.7 mm2 ). LOCE was observed in 16/288 (5.6%), 15/265 (5.7%), and 8/266 (3.0%) (P = 0.266). Target lesion revascularization (TLR) was observed in 16/288(5.6%), 12/265 (4.5%), and 4/266 (1.5%) (P = 0.042). The multivariate analysis demonstrated that smaller post-procedural MSA, as well as creatinine clearance, history of previous stroke, chronic total occlusion, and lesion SYNTAX Score was an independent predictor of TLR. In the SYNTAX II trial, larger post-procedural MSA was independently associated with the lower rate of TLR at 2 years.
Product Description
Supelco
N-Methyl-N-trimethylsilylacetamide, for GC derivatization, LiChropur, ≥97.0% (GC)