All-Aqueous Thin-Film-Flow-Induced Cell-Based Monolayers.
ACS applied materials & interfaces (2019-05-31)
Yau Kei Chan, Wing Huen Yan, Lap Tak Hung, Youchuang Chao, Jing Wu, Ho Cheung Shum
ABSTRACT
Cells in vitro usually require a solid scaffold to attach and form two-dimensional monolayer structures. To obtain a substrate-free cell monolayer, long culture time and specific detaching procedures are required. In this study, a thin-film-flow-induced strategy is reported to overcome the challenges of assembling in vitro scaffold-free monolayered cell aggregates. The assembly is driven by a dewetting-like thin-film withdrawal along all-aqueous interfaces characterized by a low interfacial tension. The withdrawal process drives the cells adsorbed on the liquid film to aggregate and assemble into an organized and compact monolayer. This strategy is not limited to biological cells but also colloidal particles, as demonstrated by the assembly of hybrid cell-particle monolayers. The versatility offered by this approach suggests new opportunities in understanding early tissue formation and functionalizing cell monolayer aggregates by colloidal particles with customized functions.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Micro particles based on polystyrene, magnetic, 10 μm particle size, std dev <0.5 μm
Supelco
Micro particles based on polystyrene, dark blue, size: 10 μm
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.