  • Search Results
  • Hapten Synthesis, Antibody Development, and a Highly Sensitive Indirect Competitive Chemiluminescent Enzyme Immunoassay for Detection of Dicamba.
Hapten Synthesis, Antibody Development, and a Highly Sensitive Indirect Competitive Chemiluminescent Enzyme Immunoassay for Detection of Dicamba.
Journal of agricultural and food chemistry (2019-05-02)
Jingqian Huo, Bogdan Barnych, Zhenfeng Li, Debin Wan, Dongyang Li, Natalia Vasylieva, Stevan Z Knezevic, O Adewale Osipitan, Jon E Scott, Jinlin Zhang, Bruce D Hammock
ABSTRACT
Although dicamba has long been one of the most widely used selective herbicides, some U.S. states have banned the sale and use of dicamba because of farmers complaints of drift and damage to nonresistant crops. To prevent illegal use of dicamba and allow monitoring of nonresistant crops, a rapid and sensitive method for detection of dicamba is critical. In this paper, three novel dicamba haptens with an aldehyde group were synthesized, conjugated to the carrier protein via a reductive-amination procedure and an indirect competitive chemiluminescent enzyme immunoassay (CLEIA) for dicamba was developed. The assay showed an IC50 of 0.874 ng/mL which was over 15 times lower than that of the conventional enzyme immunoassay. The immunoassay was used to quantify dicamba concentrations in field samples of soil and soybean obtained from fields sprayed with dicamba. The developed CLEIA showed an excellent correlation with LC-MS analysis in spike-and-recovery studies, as well as in real samples. The recovery of dicamba ranged from 86 to 108% in plant samples and from 105 to 107% in soil samples. Thus, this assay is a rapid and simple analytical tool for detecting and quantifying dicamba levels in environmental samples and potentially a great tool for on-site crop and field monitoring.
Product Description
Supelco
Clopyralid, PESTANAL®, analytical standard
Supelco
Chlorfenac, PESTANAL®, analytical standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.