  • Search Results
  • Synthesis and photodynamic activities of integrin-targeting silicon(IV) phthalocyanine-cRGD conjugates.
Synthesis and photodynamic activities of integrin-targeting silicon(IV) phthalocyanine-cRGD conjugates.
European journal of medicinal chemistry (2018-06-01)
Bi-Yuan Zheng, Xiao-Qing Yang, Yang Zhao, Qiao-Feng Zheng, Mei-Rong Ke, Tao Lin, Ren-Xun Chen, Kitty K K Ho, Naresh Kumar, Jian-Dong Huang
ABSTRACT
A series of novel symmetric or unsymmetric silicon (IV) phthalocyanines axially substituted with cyclic Arg-Gly-Asp (cRGD) ligands through different ethylene glycol chains have been synthesized by a facile and mild "click" reaction. All the compounds show efficient photosensitizing activities in N,N-dimethylformamide, and are essentially non-aggregated in RPMI 1640 medium with 0.05% Cremophor EL. Owing to the presence of two cRGD ligands, the conjugate 6b exhibits highest selectivity toward αvβ3+ HT-29 cells in photocytotoxicities. It shows higher cellular uptake and ROS generation efficiency toward the αvβ3+ HT-29 cells compared with that of αvβ3- MCF-7 cells. The competitive cellular uptake and subcellular localization indicate that 6b is internalized mainly through integrin-mediated endocytosis. In addition, the in vivo studies showed that 6b can mainly accumulate in tumor sites and show a significant PDT effect resulting in 75% tumor growth inhibition. The results indicate that 6b is a highly promising photosensitizer for targeted photodynamic therapy.
Sigma-Aldrich
Tetraethylene glycol, 99%
Sigma-Aldrich
29H,31H-Phthalocyanine, β-form, 98%