US
EN
Applications
Products
Services
Support
Home
Search Results
Ecophysiological and metabolic responses to interactive exposure to nutrients and copper excess in the brown macroalga Cystoseira tamariscifolia.
Ecophysiological and metabolic responses to interactive exposure to nutrients and copper excess in the brown macroalga Cystoseira tamariscifolia.
Marine Pollution Bulletin (2018)
Celis-Pla MSP, et al.