  • Search Results
  • Enhanced response to mouse thyroid-stimulating hormone (TSH) receptor immunization in TSH receptor-knockout mice.
Enhanced response to mouse thyroid-stimulating hormone (TSH) receptor immunization in TSH receptor-knockout mice.
Endocrinology (2010-06-18)
Mami Nakahara, Norisato Mitsutake, Hikaru Sakamoto, Chun-Rong Chen, Basil Rapoport, Sandra M McLachlan, Yuji Nagayama
ABSTRACT
Graves-like hyperthyroidism is induced in BALB/c mice by immunization with adenovirus expressing the human TSH receptor (TSHR) A-subunit (amino acids 1-289). However, because of nonidentity between the human and mouse TSHR ( approximately 87% amino acid homology), we compared the responses of mice immunized with adenoviruses expressing either the mouse or the human TSHR A-subunit. Wild-type (wt) BALB/c mice immunized with the mouse A-subunit developed neither TSHR antibodies (measured by flow cytometry) nor thyroid lymphocytic infiltration. However, wt C57BL/6 mice developed sparse intrathyroidal lymphocyte infiltration without antibody production. Depletion of naturally occurring regulatory CD4(+)CD25(+) T cells had little effect. These results indicate the inability to break tolerance to the mouse TSHR in wt mice. In contrast, TSHR knockout (KO) BALB/c mice generated mouse TSHR antibodies in response to mouse A-subunit immunization and augmented human TSHR antibody response to human A-subunit immunization. Thyroid-stimulating antibody titers measured in a functional bioassay were comparable in human A-subunit immunized wt mice and in TSHR KO mice immunized with either the mouse or human A-subunit. In conclusion, immune response to the mouse TSHR is readily induced in TSHR KO but not in wt mice. Only in the former does immunization with adenovirus expressing the mouse A-subunit generate antibodies capable of activating the mouse TSHR. TSHR KO mice are, therefore, of value for future studies dissecting the autoimmune response to the mouse TSHR.
Sigma-Aldrich
Anti-Mouse IgG (Fc specific) F(ab′)2 fragment−FITC antibody produced in goat, flow cytometry grade, affinity isolated antibody, buffered aqueous solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.