  • Search Results
  • Hantavirus-like particles generated in CHO cells induce specific immune responses in C57BL/6 mice.
Hantavirus-like particles generated in CHO cells induce specific immune responses in C57BL/6 mice.
Vaccine (2010-05-04)
Chuan Li, Feng Liu, Mifang Liang, Quanfu Zhang, Xiaofang Wang, Tao Wang, Jiandong Li, Dexin Li
ABSTRACT
A safe and effective hantavirus vaccine is highly desirable since hantaviruses are distributed worldwide and cause an acute and often fatal disease (hemorrhagic fever with renal syndrome, HFRS). Virus-like particles (VLPs) displaying functional viral proteins could provide effective vaccines against a few viruses, but their ability to induce hantavirus-specific immune response has not been adequately investigated. To measure the immunogenicity of Hantaan virus-like particles (HTN-VLPs) vaccine, we generated recombinant HTN-VLPs by co-expressing Hantaan virus nucleocapsid (N) protein and glycoproteins (Gn and Gc) in Chinese hamster ovary (CHO) cells. We compared intramuscular versus subcutaneous administration of HTN-VLPs for the ability to induce specific immune response against Hantaan virus infection. Mice that received both intramuscular and subcutaneous immunizations of HTN-VLPs were sufficiently stimulated specific antibody response against Hantaan virus N protein and glycoproteins, which was comparable to Chinese commercial inactivated bivalent hantaviruses vaccine. Moreover, vaccination with HTN-VLPs also resulted in the induction of higher levels of specific cellular response to N protein than that of inactivated vaccine. Our results provide an important insight towards the development of hantaviruses-like particles as a potential candidate vaccine for the control and prevention of hantaviruses infection.
Sigma-Aldrich
Anti-Human IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.