  • Search Results
  • A human immune dysregulation syndrome characterized by severe hyperinflammation with a homozygous nonsense Roquin-1 mutation.
A human immune dysregulation syndrome characterized by severe hyperinflammation with a homozygous nonsense Roquin-1 mutation.
Nature communications (2019-10-23)
S J Tavernier, V Athanasopoulos, P Verloo, G Behrens, J Staal, D J Bogaert, L Naesens, M De Bruyne, S Van Gassen, E Parthoens, J Ellyard, J Cappello, L X Morris, H Van Gorp, G Van Isterdael, Y Saeys, M Lamkanfi, P Schelstraete, J Dehoorne, V Bordon, R Van Coster, B N Lambrecht, B Menten, R Beyaert, C G Vinuesa, V Heissmeyer, M Dullaers, F Haerynck
ABSTRACT
Hyperinflammatory syndromes are life-threatening disorders caused by overzealous immune cell activation and cytokine release, often resulting from defects in negative feedback mechanisms. In the quintessential hyperinflammatory syndrome familial hemophagocytic lymphohistiocytosis (HLH), inborn errors of cytotoxicity result in effector cell accumulation, immune dysregulation and, if untreated, tissue damage and death. Here, we describe a human case with a homozygous nonsense R688* RC3H1 mutation suffering from hyperinflammation, presenting as relapsing HLH. RC3H1 encodes Roquin-1, a posttranscriptional repressor of immune-regulatory proteins such as ICOS, OX40 and TNF. Comparing the R688* variant with the murine M199R variant reveals a phenotypic resemblance, both in immune cell activation, hypercytokinemia and disease development. Mechanistically, R688* Roquin-1 fails to localize to P-bodies and interact with the CCR4-NOT deadenylation complex, impeding mRNA decay and dysregulating cytokine production. The results from this unique case suggest that impaired Roquin-1 function provokes hyperinflammation by a failure to quench immune activation.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Sodium (meta)arsenite, ≥90%
Sigma-Aldrich
Ionomycin calcium salt, Ready Made Solution, from Streptomyces conglobatus, 1 mM in DMSO, 0.2 μm filtered
Sigma-Aldrich
Pyridinium chlorochromate, 98%
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
BIO, ≥98% (HPLC)
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%