  • Search Results
  • Amphiphilic Polymethyloxazoline-Polyethyleneimine Copolymers: Interaction with Lipid Bilayer and Antibacterial Properties.
Amphiphilic Polymethyloxazoline-Polyethyleneimine Copolymers: Interaction with Lipid Bilayer and Antibacterial Properties.
Macromolecular bioscience (2019-11-21)
Dominika Kozon, Jolanta Mierzejewska, Tomasz Kobiela, Agnieszka Grochowska, Ksenia Dudnyk, Agnieszka Głogowska, Anna Sobiepanek, Aleksandra Kuźmińska, Tomasz Ciach, Ewa Augustynowicz-Kopeć, Dominik Jańczewski
ABSTRACT
Polycations, mimicking activity of antibacterial peptides, belong to an important class of molecules investigated as a support or as an alternative to antibiotics. In this work, studies of modified linear amphiphilic statistical polymethyloxazoline (PMOX) and polyethyleneimine copolymers (PMOX_PEI) series are presented. Variation of PEI content in the structure results in controllable changes of polymeric aggregates zeta potential. The structure with the highest positive charge shows the best antimicrobial activity, well visible in tests against model Gram-positive and Gram-negative bacteria, fungi, and mycobacterium strains. The polymer toxicity is evaluated with MTT and hemolysis assay as a reference. Quartz crystal microbalance (QCM-D) is used to investigate interaction between polycations and a model lipid membrane. Polymer activity correlates well with molecular structure, showing that amphiphilic component is altering polymer behavior in contact with the lipid bilayer.
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
2-Methyl-2-oxazoline, 98%
Sigma-Aldrich
Magnesium citrate, AldrichCPR
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.